Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937951.3 | XP_004938008.1 | 4 - 5 | 638 | 70 | |
XM_015286371.2 | XP_015141857.1 | 4 - 5 | 581 | 70 | ||
XM_015286377.2 | XP_015141863.1 | 4 - 5 | 639 | 70 | ||
XM_416394.6 | XP_416394.1 | 4 - 5 | 630 | 70 | ||
GenBank mRNA | CR353237.1 | - | 2 - 3 | 236 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |