Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281969.1 | XP_015137455.1 | 3 - 4 | 124 | ||
GenBank mRNA | CR353752.1 | - | 3 - 4 | 339 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |