Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277559.1 | NP_001264488.1 | 9 - 10 | 1188 | 76 | |
XM_015294202.2 | XP_015149688.1 | 9 - 10 | 1223 | 76 | ||
XM_015294203.2 | XP_015149689.1 | 8 - 9 | 1103 | 76 | ||
GenBank mRNA | BX929329.2 | - | 9 - 10 | 1189 | 76 | |
BX935844.1 | - | 9 - 10 | 1189 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |