Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930455.2 | - | 8 - 9 | 1193 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |