Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199587.1 | NP_001186516.1 | 3 - 4 | 550 | 133 | |
GenBank mRNA | BX932088.2 | - | 3 - 4 | 476 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |