Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199598.1 | NP_001186527.1 | 2 - 3 | 403 | 129 | |
GenBank mRNA | BX932258.2 | - | 2 - 3 | 118 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |