Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419766.6 | XP_419766.4 | 2 - 3 | 694 | 88 | |
GenBank mRNA | BX933225.2 | - | 2 - 3 | 376 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |