Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419699.5 | XP_419699.1 | 1 - 2 | 100 | 90 | |
GenBank mRNA | BX929526.1 | - | 1 - 2 | 90 | ||
BX933267.2 | - | 1 - 2 | 73 | 90 | ||
BX934469.1 | - | 1 - 2 | 85 | 90 | ||
CR353789.1 | - | 1 - 2 | 42 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |