Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277819.1 | NP_001264748.1 | 3 - 4 | 341 | 77 | |
XM_015285433.2 | XP_015140919.1 | 4 - 5 | 374 | 77 | ||
XM_025149683.1 | XP_025005451.1 | 4 - 5 | 661 | 77 | ||
XM_025149684.1 | XP_025005452.1 | 4 - 5 | 620 | 77 | ||
GenBank mRNA | BU325679.1 | - | 3 - 4 | 319 | 77 | |
BX933437.2 | - | 3 - 4 | 341 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |