Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015286685.2 | XP_015142171.1 | 8 - 9 | 1187 | 98 | |
GenBank mRNA | BX934822.2 | - | 8 - 9 | 1127 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |