Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTHRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418373.6 | XP_418373.5 | 4 - 5 | 1122 | 75 | |
GenBank mRNA | BX935556.2 | - | 3 - 4 | 577 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |