Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297915.2 | XP_015153401.1 | 3 - 4 | 811 | 127 | |
XM_025143141.1 | XP_024998909.1 | 3 - 4 | 811 | 127 | ||
XM_025143142.1 | XP_024998910.1 | 3 - 4 | 542 | 127 | ||
XM_025143143.1 | XP_024998911.1 | 3 - 4 | 811 | 127 | ||
XM_025143144.1 | XP_024998912.1 | 3 - 4 | 811 | 127 | ||
GenBank mRNA | BX935722.2 | - | 4 - 5 | 703 | 127 | |
FJ947118.1 | - | 3 - 4 | 367 | 127 | ||
FJ947119.1 | - | 3 - 4 | 367 | 127 |
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KF680104.1 | - | 3 - 4 | 367 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |