Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252611.1 | NP_001239540.1 | 1 - 2 | 411 | 143 | |
GenBank mRNA | BM490904.1 | - | 1 - 2 | 412 | 144 | |
CO422524.1 | - | 1 - 2 | 134 | 143 | ||
CR353783.1 | - | 1 - 2 | 127 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |