Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252285.1 | NP_001239214.1 | 5 - 6 | 722 | 134 | |
GenBank mRNA | CR353823.1 | - | 4 - 5 | 515 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |