Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199565.1 | NP_001186494.1 | 7 - 8 | 687 | 110 | |
XM_025150660.1 | XP_025006428.1 | 7 - 8 | 1208 | 110 | ||
XM_025150664.1 | XP_025006432.1 | 7 - 8 | 637 | 110 | ||
GenBank mRNA | BU253847.1 | - | 5 - 6 | 435 | 110 | |
BX929358.2 | - | 7 - 8 | 680 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |