Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199711.1 | NP_001186640.1 | 2 - 3 | 314 | 84 | |
XM_015287352.2 | XP_015142838.1 | 3 - 4 | 416 | 84 | ||
XM_015287353.2 | XP_015142839.1 | 3 - 4 | 335 | 84 | ||
XM_015287354.2 | XP_015142840.1 | 3 - 4 | 418 | 84 | ||
XM_025150736.1 | XP_025006504.1 | 3 - 4 | 501 | 84 | ||
GenBank mRNA | BX929546.2 | - | 3 - 4 | 410 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |