Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280453.2 | XP_015135939.1 | 11 - 12 | 3291 | 84 | |
XM_015280465.2 | XP_015135951.1 | 12 - 13 | 1572 | 84 | ||
XM_015280475.2 | XP_015135961.1 | 11 - 12 | 1631 | 84 | ||
XM_025146897.1 | XP_025002665.1 | 13 - 14 | 1721 | 84 | ||
XM_417196.6 | XP_417196.1 | 12 - 13 | 1616 | 84 | ||
GenBank mRNA | BX929784.2 | - | 10 - 11 | 1015 | 84 | |
CR353417.1 | - | 1 - 2 | 845 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |