Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280465.2 | XP_015135951.1 | 7 - 8 | 1285 | 86 | |
XM_015280475.2 | XP_015135961.1 | 7 - 8 | 1402 | 86 | ||
XM_417196.6 | XP_417196.1 | 7 - 8 | 1329 | 86 | ||
GenBank mRNA | BX929784.2 | - | 5 - 6 | 728 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |