Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX931443.2 | - | 1 - 2 | 379 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |