Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415108.5 | XP_415108.2 | 20 - 21 | 69 | ||
GenBank mRNA | BX933081.2 | - | 1 - 2 | 84 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |