Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277592.1 | NP_001264521.1 | 5 - 6 | 939 | 75 | |
GenBank mRNA | BX933119.2 | - | 5 - 6 | 928 | 75 | |
CR406559.1 | - | 4 - 5 | 527 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |