Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416745.6 | XP_416745.1 | 1 - 2 | 83 | 103 | |
GenBank mRNA | BX933509.2 | - | 1 - 2 | 71 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |