Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146831.1 | XP_025002599.1 | 3 - 4 | 579 | 87 | |
XM_417110.6 | XP_417110.2 | 3 - 4 | 634 | 87 | ||
GenBank mRNA | BX933581.2 | - | 3 - 4 | 352 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |