Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX950354.2 | - | 15 - 16 | 1608 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |