Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001349739.1 | NP_001336668.1 | 1 - 2 | 67 | 84 | |
GenBank mRNA | BX950579.2 | - | 1 - 2 | 73 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |