Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199526.1 | NP_001186455.1 | 1 - 2 | 59 | 93 | |
XM_015288010.2 | XP_015143496.1 | 1 - 2 | 59 | 93 | ||
GenBank mRNA | BU411926.1 | - | 1 - 2 | 43 | 93 | |
CR354339.1 | - | 1 - 2 | 59 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |