Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199526.1 | NP_001186455.1 | 3 - 4 | 159 | 105 | |
XM_025151511.1 | XP_025007279.1 | 3 - 4 | 173 | 105 | ||
GenBank mRNA | BU411926.1 | - | 3 - 4 | 143 | 105 | |
CR354339.1 | - | 3 - 4 | 159 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |