Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277755.1 | NP_001264684.1 | 2 - 3 | 166 | 112 | |
GenBank mRNA | CO635620.1 | - | 1 - 2 | 133 | 112 | |
CR387290.1 | - | 1 - 2 | 133 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |