Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416786.6 | XP_416786.1 | 1 - 2 | 345 | 92 | |
GenBank mRNA | CR388761.1 | - | 1 - 2 | 329 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |