Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276813.2 | XP_015132299.1 | 2 | 3240 | 95 | |
XM_015276818.2 | XP_015132304.1 | 2 | 3167 | 95 | ||
XM_015276826.2 | XP_015132312.1 | 1 | 3673 | 95 | ||
XM_015276837.2 | XP_015132323.1 | 1 | 3673 | 95 | ||
GenBank mRNA | AF098476.1 | - | 1 | 351 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |