Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641442.4 | XP_003641490.2 | 1 - 2 | 852 | 113 | |
XM_015287909.2 | XP_015143395.2 | 1 - 2 | 852 | 113 | ||
GenBank mRNA | AJ133778.1 | - | 1 - 2 | 465 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |