Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277440.1 | NP_001264369.1 | 3 - 4 | 421 | 132 | |
NM_001277441.1 | NP_001264370.1 | 3 - 4 | 383 | 132 | ||
NM_001277442.1 | NP_001264371.1 | 2 - 3 | 318 | 132 | ||
XM_015276926.2 | XP_015132412.1 | 3 - 4 | 496 | 132 | ||
XM_015276927.2 | XP_015132413.1 | 3 - 4 | 279 | 132 | ||
XM_015276928.2 | XP_015132414.1 | 3 - 4 | 314 | 132 | ||
XM_025153355.1 | XP_025009123.1 | 3 - 4 | 460 | 132 | ||
XM_025153356.1 | XP_025009124.1 | 3 - 4 | 355 | 132 | ||
GenBank mRNA | BX929424.1 | - | 3 - 4 | 383 | 132 | |
CN229900.1 | - | 3 - 4 | 338 | 132 | ||
CN237805.1 | - | 3 - 4 | 421 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |