Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277556.3 | NP_001264485.2 | 1 - 2 | 92 | 112 | |
XM_025152827.1 | XP_025008595.1 | 1 - 2 | 125 | 112 | ||
GenBank mRNA | BU109727.1 | - | 1 - 2 | 92 | 112 | |
BX930646.1 | - | 1 - 2 | 90 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |