Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329508.1 | NP_001316437.1 | 8 - 9 | 841 | 76 | |
XM_015294357.2 | XP_015149843.1 | 8 - 9 | 892 | 76 | ||
GenBank mRNA | BX931257.1 | - | 8 - 9 | 842 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |