Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277490.1 | NP_001264419.1 | 6 - 7 | 689 | 77 | |
GenBank mRNA | BX932848.1 | - | 6 - 7 | 660 | 77 | |
DR424316.1 | - | 7 - 8 | 637 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |