Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294453.2 | XP_015149939.1 | 2 - 3 | 336 | 78 | |
XM_414851.6 | XP_414851.3 | 2 - 3 | 598 | 78 | ||
GenBank mRNA | BX933578.1 | - | 252 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |