Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941851.3 | XP_004941908.1 | 2 - 3 | 125 | ||
XM_421305.6 | XP_421305.1 | 2 - 3 | 125 | |||
GenBank mRNA | BX933733.1 | - | 2 - 3 | 200 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |