Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416293.6 | XP_416293.3 | 3 | 3080 | 116 | |
GenBank mRNA | BX933905.1 | - | 1 | 2047 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |