Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299083.2 | XP_015154569.1 | 1280 | 60 | ||
XM_015299084.2 | XP_015154570.1 | 1287 | 60 | |||
XM_015299085.2 | XP_015154571.1 | 1382 | 60 | |||
XM_015299086.2 | XP_015154572.1 | 1037 | 60 | |||
XM_417963.6 | XP_417963.3 | 1257 | 60 | |||
GenBank mRNA | BX935746.1 | - | 688 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus |