Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277413.1 | NP_001264342.1 | 11 - 12 | 917 | 138 | |
XM_025147097.1 | XP_025002865.1 | 12 - 13 | 1153 | 138 | ||
XM_025147098.1 | XP_025002866.1 | 11 - 12 | 1096 | 138 | ||
XM_025147099.1 | XP_025002867.1 | 12 - 13 | 1030 | 138 | ||
XM_025147100.1 | XP_025002868.1 | 11 - 12 | 1108 | 138 | ||
XM_025147101.1 | XP_025002869.1 | 10 - 11 | 969 | 138 | ||
XM_025147102.1 | XP_025002870.1 | 10 - 11 | 1054 | 138 | ||
XM_025147103.1 | XP_025002871.1 | 10 - 11 | 887 | 138 | ||
GenBank mRNA | BX936064.1 | - | 11 - 12 | 916 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |