Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271894.1 | NP_001258823.1 | 7 - 8 | 1574 | 60 | |
GenBank mRNA | Y13978.1 | - | 1584 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |