Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205329.1 | NP_990660.1 | 2 - 3 | 210 | 77 | |
GenBank mRNA | Z21720.1 | - | 210 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |