Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012932.2 | NP_001012950.2 | 4 - 5 | 626 | 71 | |
XM_015298619.2 | XP_015154105.2 | 4 - 5 | 887 | 71 | ||
GenBank mRNA | AJ720606.1 | - | 4 - 5 | 626 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |