Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ851434.1 | - | 6 - 7 | 788 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |