Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276787.2 | XP_015132273.1 | 3 - 4 | 1399 | 109 | |
GenBank mRNA | CR391481.1 | - | 1 - 2 | 314 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |