Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935711.3 | XP_004935768.2 | 3 - 4 | 365 | 145 | |
XM_004935712.2 | XP_004935769.2 | 2 - 3 | 232 | 145 | ||
XM_004935713.3 | XP_004935770.2 | 2 - 3 | 225 | 145 | ||
XM_015284178.2 | XP_015139664.1 | 3 - 4 | 364 | 145 | ||
XM_015284179.2 | XP_015139665.1 | 2 - 3 | 196 | 145 | ||
XM_015284180.2 | XP_015139666.1 | 3 - 4 | 366 | 145 | ||
XM_025149344.1 | XP_025005112.1 | 2 - 3 | 135 | 145 | ||
GenBank mRNA | CR406255.1 | - | 2 - 3 | 237 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |