Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX932894.1 | - | 2 | 456 | 156 | |
BX934270.2 | - | 2 | 452 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |