Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006313.2 | NP_001006313.2 | 7 - 8 | 1313 | 59 | |
XM_015297665.2 | XP_015153151.1 | 8 - 9 | 1352 | 59 | ||
XM_015297666.2 | XP_015153152.1 | 8 - 9 | 1906 | 59 | ||
XM_025142994.1 | XP_024998762.1 | 8 - 9 | 1331 | 59 | ||
XM_025142995.1 | XP_024998763.1 | 9 - 10 | 1437 | 59 | ||
GenBank mRNA | AJ720022.1 | - | 8 - 9 | 1353 | 59 | |
CR352474.1 | - | 1 - 2 | 476 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |