Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006334.1 | NP_001006334.1 | 3 - 4 | 432 | 117 | |
XM_015299602.2 | XP_015155088.1 | 4 - 5 | 676 | 117 | ||
XM_015299603.2 | XP_015155089.1 | 4 - 5 | 383 | 117 | ||
XM_015299604.2 | XP_015155090.1 | 4 - 5 | 426 | 117 | ||
XM_025143978.1 | XP_024999746.1 | 2 - 3 | 243 | 117 | ||
GenBank mRNA | AJ720128.1 | - | 3 - 4 | 432 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |