Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006335.2 | NP_001006335.2 | 9 - 10 | 925 | 108 | |
XM_015299606.2 | XP_015155092.1 | 9 - 10 | 932 | 108 | ||
GenBank mRNA | AJ719694.1 | - | 9 - 10 | 925 | 108 | |
CD761342.1 | - | 3 - 4 | 443 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |